siRNA Details
Gene Symbol: Stard3nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008298.1 | NP_001008299.1 | 6 | 617 | ||
GenBank mRNA | BC086352.1 | - | 6 | 617 | 7303026 | |
FQ220233.1 | - | 6 | 640 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_024270, AK152378, AK030365, AK002760, BG084215, and others.
