siRNA Details
Gene Symbol: Maea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008319.2 | NP_001008320.2 | 9 | 1907 | ||
GenBank mRNA | BC085770.1 | - | 9 | 1878 | 7189726 | |
FQ224122.1 | - | 1 | 505 |

More Information
Other Species Targeted

Gene Ontology Categories:
