siRNA Details
Gene Symbol: LCMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014793.4 | NP_055608.2 | 1 | 1122 | ||
GenBank mRNA | AF265443.1 | - | 1 | 615 | ||
AK292587.1 | - | 1 | 1048 | |||
AK300820.1 | - | 2 | 331 | |||
BC015949.2 | - | 1 | 1105 | 3922888 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
