siRNA Details
Gene Symbol: Snx24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008364.1 | NP_001008365.1 | 6, 7 | 853 | ||
XM_008772152.1 | XP_008770374.1 | 7, 8 | 900 | |||
GenBank mRNA | BC085883.1 | - | 6, 7 | 853 | 7192093 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029394, AK017610, BC051168, AK012711.
Gene Ontology Categories:
