siRNA Details
Gene Symbol: Pitpnm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008369.1 | NP_001008370.1 | 24 | 3946 | ||
GenBank mRNA | BC085945.1 | - | 24 | 3946 | 7128360 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
