siRNA Details
Gene Symbol: Sdhaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008371.1 | NP_001008372.1 | 4 | 431 | ||
XM_006231042.1 | XP_006231104.1 | 4 | 528 | |||
XM_006231043.2 | XP_006231105.1 | 5 | 545 | |||
GenBank mRNA | BC076396.1 | - | 5 | 443 | ||
BC086542.1 | - | 4 | 431 | 7122954 |
