siRNA Details
Gene Symbol: Lamtor3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008375.1 | NP_001008376.1 | 5 | 312 | ||
XM_006233383.2 | XP_006233445.1 | 5 | 437 | |||
XM_006233385.2 | XP_006233447.1 | 5 | 382 | |||
GenBank mRNA | BC086353.1 | - | 5 | 312 | 7304027 |
