siRNA Details
Gene Symbol: Prr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008379.1 | NP_001008380.1 | 3 | 382 | ||
XM_008765769.1 | XP_008763991.1 | 3 | 459 | |||
GenBank mRNA | BC086446.1 | - | 3 | 382 | 7303132 | |
FQ220513.1 | - | 3 | 365 | |||
FQ221261.1 | - | 3 | 366 | |||
FQ225143.1 | - | 3 | 366 | |||
FQ225286.1 | - | 3 | 366 | |||
FQ227365.1 | - | 3 | 366 | |||
FQ229376.1 | - | 3 | 366 | |||
FQ229733.1 | - | 3 | 366 | |||
FQ229867.1 | - | 3 | 365 | |||
FQ234745.1 | - | 3 | 403 |
