siRNA Details
Gene Symbol: Dis3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008380.1 | NP_001008381.1 | 5 | 618 | ||
XM_006243261.2 | XP_006243323.1 | 5 | 721 | |||
XM_006243262.2 | XP_006243324.1 | 5 | 525 | |||
XM_006243263.2 | XP_006243325.1 | 5 | 1050 | |||
XM_008766311.1 | XP_008764533.1 | 5 | 721 | |||
GenBank mRNA | BC085932.1 | - | 5 | 618 | 7112012 |
