siRNA Details
Gene Symbol: Thoc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008425.1 | NP_001008425.1 | 11 | 977 | ||
GenBank mRNA | AK089746.1 | - | 8 | 1305 | ||
BC085131.1 | - | 11 | 977 | 5715435 | ||
BC089551.1 | - | 12 | 977 | 30303151 |
