siRNA Details
Gene Symbol: Gm5595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008427.1 | NP_001008427.1 | 4 | 488 | ||
GenBank mRNA | BC068175.1 | - | 4 | 488 | 30550329 |
Gene Symbol: EU599041 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177525.1 | NP_001170996.1 | 4 | 529 | ||
GenBank mRNA | EU599041.1 | - | 5 | 593 | ||
EU599042.1 | - | 4 | 529 |
