siRNA Details
Gene Symbol: PHOSPHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008489.3 | NP_001008489.1 | 4 | 786 | ||
NM_001199285.1 | NP_001186214.1 | 4 | 686 | |||
NM_001199286.1 | NP_001186215.1 | 4 | 905 | |||
NM_001199287.1 | NP_001186216.1 | 5 | 904 | |||
NM_001199288.1 | NP_001186217.1 | 4 | 705 | |||
GenBank mRNA | AK314915.1 | - | 4 | 689 | ||
BC022324.2 | - | 3 | 617 | 4701753 | ||
BC106013.1 | - | 4 | 775 | 4387304 |
