siRNA Details
Gene Symbol: TMCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008740.3 | NP_001008740.1 | 2 | 423 | ||
GenBank mRNA | BC108666.1 | - | 2 | 356 | 5743890 | |
BU567634.1 | - | 2 | 426 | |||
BU569147.1 | - | 2 | 392 | |||
DB518323.1 | - | 2 | 124 |
