siRNA Details
Gene Symbol: Wdr77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008771.3 | NP_001008771.2 | 7 | 773 | ||
GenBank mRNA | AY321348.1 | - | 8 | 734 | ||
BC097367.1 | - | 7 | 724 | 7443204 | ||
FQ213175.1 | - | 7 | 868 |
Gene Symbol: LOC100911453 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749373.3 | XP_003749421.1 | 7 | 879 |
