siRNA Details
Gene Symbol: FBXO47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008777.2 | NP_001008777.2 | 9 | 1269 | ||
XM_011524865.2 | XP_011523167.1 | 9 | 1264 | |||
XM_011524866.2 | XP_011523168.1 | 8 | 1171 | |||
XM_011524867.2 | XP_011523169.1 | 9 | 1342 | |||
GenBank mRNA | AY700575.1 | - | 10 | 1269 | ||
BC140879.1 | - | 10 | 1239 | 9021747 | ||
BC140880.1 | - | 10 | 1239 | 9021748 |
