siRNA Details
Gene Symbol: KIAA0101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014736.5 | NP_055551.1 | 3 | 406 | ||
NR_109934.1 | 3 | 462 | ||||
GenBank mRNA | AF529370.1 | - | 3 | 267 | ||
AK290748.1 | - | 3 | 337 | |||
AY598324.1 | - | 3 | 267 | |||
BC005832.2 | - | 3 | 326 | 2988995 | ||
BC007101.1 | - | 3 | 333 | 4281110 | ||
BC016782.1 | - | 3 | 341 | 4109322 | ||
BF691807.1 | - | 3 | 407 | |||
BG502841.1 | - | 399 | ||||
D14657.1 | - | 3 | 328 | |||
DT220867.1 | - | 3 | 459 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
