siRNA Details
Gene Symbol: Ift122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001057925.4 | XP_001057925.2 | 14 | 1754 | ||
XM_006237067.1 | XP_006237129.1 | 15 | 1931 | |||
GenBank mRNA | BC087667.1 | - | 14 | 1604 | 7317105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
