siRNA Details
Gene Symbol: Oas1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009490.1 | NP_001009490.1 | 6 | 1442 | ||
GenBank mRNA | AY196700.1 | - | 6 | 1442 |
Gene Symbol: LOC100910735 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751197.2 | XP_003751245.1 | 1656 |
Gene Symbol: Oas1i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009680.1 | NP_001009680.1 | 6 | 1636 | ||
GenBank mRNA | AY196697.1 | - | 6 | 1636 | ||
BC160819.1 | - | 6 | 1610 | 7382468 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
