siRNA Details
Gene Symbol: Eogt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009502.1 | NP_001009502.1 | 11, 12 | 1396 | ||
XM_008763144.1 | XP_008761366.1 | 8, 9 | 1070 | |||
XM_008763145.1 | XP_008761367.1 | 9, 10 | 982 | |||
XM_008763146.1 | XP_008761368.1 | 7, 8 | 862 | |||
XM_008763147.1 | XP_008761369.1 | 8, 9 | 1127 | |||
GenBank mRNA | AJ868236.1 | - | 11, 12 | 1396 |
