siRNA Details
Gene Symbol: LOC494224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009505.2 | NP_001009505.1 | 3, 4 | 132 | ||
XM_006250794.2 | XP_006250856.1 | 2, 3 | 273 | |||
GenBank mRNA | AB196306.1 | - | 3, 4 | 132 |

More Information
Other Species Targeted

