siRNA Details
Gene Symbol: RUSC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135999.1 | NP_001129471.1 | 4 | 3315 | ||
NM_014806.3 | NP_055621.1 | 4 | 3059 | |||
NR_052015.2 | 4 | 2734 | ||||
XM_006716896.2 | XP_006716959.1 | 3 | 2906 | |||
GenBank mRNA | AB002373.2 | - | 4 | 2901 | ||
AK299805.1 | - | 4 | 3005 | |||
BC064843.1 | - | 3 | 2103 | |||
BC132766.1 | - | 3 | 2844 | 40146788 | ||
BC132770.1 | - | 3 | 2844 | 40146792 | ||
BC146654.1 | - | 4 | 3033 | 40147826 | ||
BC150211.1 | - | 4 | 2646 | |||
BC150262.1 | - | 4 | 2912 | 9007205 |
