siRNA Details
Gene Symbol: TMEM63B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318792.1 | NP_001305721.1 | 18, 19 | 1851 | ||
NM_018426.2 | NP_060896.1 | 18, 19 | 1817 | |||
XM_005249211.3 | XP_005249268.1 | 20, 21 | 2197 | |||
XM_005249213.4 | XP_005249270.1 | 19, 20 | 2124 | |||
XM_005249214.4 | XP_005249271.1 | 17, 18 | 1711 | |||
XM_005249215.4 | XP_005249272.1 | 18, 19 | 2016 | |||
XM_005249217.1 | XP_005249274.1 | 18, 19 | 1889 | |||
XM_006715135.1 | XP_006715198.1 | 18, 19 | 1873 | |||
XM_017010999.1 | XP_016866488.1 | 20, 21 | 2074 | |||
XM_017011000.1 | XP_016866489.1 | 19, 20 | 2055 | |||
XM_017011001.1 | XP_016866490.1 | 17, 18 | 1755 | |||
XM_017011002.1 | XP_016866491.1 | 17, 18 | 1858 | |||
GenBank mRNA | AL353957.1 | - | 4, 5 | 275 | ||
AL834238.1 | - | 11, 12 | 1115 | |||
BC022095.1 | - | 15, 16 | 1454 | |||
BC062989.1 | - | 6, 7 | 643 | |||
BC136769.1 | - | 17, 18 | 1693 | 9020759 | ||
CR627460.1 | - | 18, 19 | 1854 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_198167, BC059283, BC039277, BC026370, AK090200, and others.
Gene Ontology Categories:
