siRNA Details
Gene Symbol: TMEM63C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020431.3 | NP_065164.2 | 24 | 3439 | ||
GenBank mRNA | AK124749.1 | - | 10 | 1947 | ||
AK125159.1 | - | 24 | 3371 | |||
AL137278.1 | - | 1 | 62 | |||
AL157492.1 | - | 5 | 1392 | |||
BC136613.1 | - | 24 | 3321 | 9020603 | ||
BC136614.1 | - | 24 | 3318 | 9020604 |
