siRNA Details
Gene Symbol: ZYG11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024646.2 | NP_078922.1 | 3 | 1137 | ||
XM_006710898.3 | XP_006710961.1 | 3 | 1024 | |||
XM_017002336.1 | XP_016857825.1 | 3 | 1142 | |||
GenBank mRNA | AB051517.1 | - | 3 | 1087 | ||
AK056645.1 | - | 1 | 81 | |||
AK095643.1 | - | 2 | 777 | |||
AK296460.1 | - | 3 | 1142 | |||
BC029832.1 | - | 1 | 151 | |||
BX647643.1 | - | 1 | 310 | |||
DQ646393.1 | - | 3 | 1156 |
