siRNA Details
Gene Symbol: CEP290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025114.3 | NP_079390.3 | 38 | 5449 | ||
XM_011538756.2 | XP_011537058.1 | 40 | 6310 | |||
XM_011538757.2 | XP_011537059.1 | 40 | 6118 | |||
XM_011538758.2 | XP_011537060.1 | 40 | 6310 | |||
XM_011538759.2 | XP_011537061.1 | 40 | 6310 | |||
XM_011538760.1 | XP_011537062.1 | 40 | 6310 | |||
XM_011538761.1 | XP_011537063.1 | 40 | 6310 | |||
XM_011538762.2 | XP_011537064.1 | 39 | 5542 | |||
XM_011538763.2 | XP_011537065.1 | 38 | 5449 | |||
XM_011538764.2 | XP_011537066.1 | 40 | 6310 | |||
XM_011538765.2 | XP_011537067.1 | 40 | 6310 | |||
XM_011538766.2 | XP_011537068.1 | 26 | 4537 | |||
XM_017019980.1 | XP_016875469.1 | 40 | 6310 | |||
XM_017019981.1 | XP_016875470.1 | 40 | 6310 | |||
XM_017019982.1 | XP_016875471.1 | 40 | 6310 | |||
XM_017019983.1 | XP_016875472.1 | 37 | 5428 | |||
GenBank mRNA | AB002371.1 | - | 13 | 3465 | ||
AF273044.1 | - | 2 | 262 | |||
DQ109808.1 | - | 38 | 5449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
