siRNA Details
Gene Symbol: Cbx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028763.3 | NP_083039.2 | 5 | 481 | ||
GenBank mRNA | AK004679.2 | - | 5 | 386 | ||
AK035250.1 | - | 5 | 388 | |||
AK090253.1 | - | 5 | 333 | |||
AK146048.1 | - | 5 | 332 | |||
AK146103.1 | - | 5 | 332 | |||
AK147702.1 | - | 1 | 5084 | |||
BC011199.1 | - | 5 | 370 | |||
BC048240.1 | - | 5 | 378 | 6493113 |
Gene Symbol: Npcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013362.2 | NP_001013380.2 | 5 | 481 | ||
GenBank mRNA | AY394489.1 | - | 5 | 386 | ||
AY395907.1 | - | 5 | 386 |

More Information
Other Species Targeted

Gene Ontology Categories:
