siRNA Details
Gene Symbol: 9530053H05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC005505.1 | - | 3 | 1614 | ||
BC049918.1 | - | 1 | 464 | |||
BC057455.1 | - | 369 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
