siRNA Details
Gene Symbol: GPRIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005277666.3 | XP_005277723.1 | 4 | 3770 | ||
XM_011540388.2 | XP_011538690.1 | 6 | 4130 | |||
XM_011540395.2 | XP_011538697.1 | 4 | 3738 | |||
XM_011540400.2 | XP_011538702.1 | 3 | 3615 | |||
XM_017016974.1 | XP_016872463.1 | 6 | 6035 | |||
XM_017016975.1 | XP_016872464.1 | 5 | 6369 | |||
XM_017016976.1 | XP_016872465.1 | 6 | 6310 | |||
XM_017016977.1 | XP_016872466.1 | 6 | 4542 | |||
XM_017016978.1 | XP_016872467.1 | 5 | 6423 | |||
XM_017016979.1 | XP_016872468.1 | 4 | 4303 | |||
XM_017016980.1 | XP_016872469.1 | 4 | 4265 | |||
XM_017016981.1 | XP_016872470.1 | 5 | 5880 | |||
XM_017016982.1 | XP_016872471.1 | 5 | 5896 | |||
XM_017016983.1 | XP_016872472.1 | 5 | 6171 | |||
XM_017016984.1 | XP_016872473.1 | 5 | 6155 | |||
GenBank mRNA | AB011086.1 | - | 1 | 4319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
