siRNA Details
Gene Symbol: Spp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053577.1 | NP_446029.1 | 738 | |||
GenBank mRNA | BC088193.1 | - | 738 | 7372900 | ||
FQ210158.1 | - | 654 | ||||
FQ210198.1 | - | 675 | ||||
FQ210350.1 | - | 702 | ||||
FQ218693.1 | - | 96 |
