siRNA Details
Gene Symbol: Ppp2r1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057140.1 | NP_476481.1 | 9 | 1023 | ||
GenBank mRNA | BC083859.1 | - | 9 | 1023 | 7126254 |

More Information
Other Species Targeted

Gene Ontology Categories:
