siRNA Details
Gene Symbol: A430105I19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001982.2 | NP_001001982.2 | 11 | 4144 | ||
GenBank mRNA | AK137584.1 | - | 11 | 4146 | ||
BC075684.1 | - | 10 | 3896 | 30611649 |
