siRNA Details
Gene Symbol: Zfp948 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002008.1 | NP_001002008.1 | 6 | 2509 | ||
GenBank mRNA | AK164108.1 | - | 6 | 2547 | ||
BC049807.1 | - | 6 | 2511 | 6333379 | ||
BC076634.1 | - | 6 | 2509 | 30659397 |
