siRNA Details
Gene Symbol: A830005F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK043525.1 | - | 2 | 249 | ||
BB262224.2 | - | 1 | 129 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A830005F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK043525.1 | - | 2 | 249 | ||
BB262224.2 | - | 1 | 129 |