siRNA Details
Gene Symbol: Higd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002900.1 | NP_001002900.1 | 4 | 308 | ||
GenBank mRNA | AB099517.1 | - | 4 | 308 | ||
AB110950.1 | - | 4 | 822 | |||
AB253942.1 | - | 4 | 348 |
Gene Symbol: Methig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024672.3 | NP_001019843.1 | 4 | 771 | ||
GenBank mRNA | AB099516.1 | - | 4 | 808 | ||
AK144032.1 | - | 4 | 772 | |||
BC139180.1 | - | 4 | 805 | 8862202 | ||
BC139182.1 | - | 4 | 805 | 8862204 | ||
BC145570.1 | - | 3 | 708 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
