siRNA Details
Gene Symbol: Dnmt3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003959.1 | NP_001003959.1 | 10, 11 | 1493 | ||
XM_006235340.2 | XP_006235402.1 | 9, 10 | 1363 | |||
XM_006235342.2 | XP_006235404.1 | 9, 10 | 1486 | |||
XM_006235347.2 | XP_006235409.1 | 8, 9 | 1301 | |||
XM_008762364.1 | XP_008760586.1 | 8, 9 | 2765 | |||
XM_008762365.1 | XP_008760587.1 | 8, 9 | 2765 | |||
XM_008762366.1 | XP_008760588.1 | 8, 9 | 2765 | |||
XM_008762368.1 | XP_008760590.1 | 9, 10 | 1143 | |||
XM_008762370.1 | XP_008760592.1 | 8, 9 | 2765 | |||
XM_008762371.1 | XP_008760593.1 | 8, 9 | 2765 |
