siRNA Details
Gene Symbol: PHYHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099335.1 | NP_001092805.1 | 6 | 1999 | ||
NM_014759.3 | NP_055574.3 | 5 | 1877 | |||
XM_017014102.1 | XP_016869591.1 | 3 | 2142 | |||
GenBank mRNA | AK098743.1 | - | 6 | 1622 | ||
AK294372.1 | - | 4 | 1329 | |||
BC034034.1 | - | 5 | 1630 | 4793002 | ||
BC035940.1 | - | 5 | 1630 | 4793228 | ||
D87463.1 | - | 6 | 1708 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
