siRNA Details
Gene Symbol: Abcg3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004076.2 | NP_001004076.2 | 5 | 485 | ||
XM_006250626.2 | XP_006250688.1 | 5 | 534 | |||
XM_008770002.1 | XP_008768224.1 | 5 | 2875 | |||
XM_008770003.1 | XP_008768225.1 | 6 | 2755 | |||
GenBank mRNA | AY688113.1 | - | 4 | 412 | ||
BC098896.1 | - | 5 | 485 | 7100556 |
Gene Symbol: Abcg3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014133.1 | NP_001014155.1 | 379 | |||
GenBank mRNA | BC085773.1 | - | 379 | 7096429 |
Gene Symbol: Abcg3l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107205.1 | NP_001100675.1 | 5 | 470 | ||
XM_006250606.2 | XP_006250668.1 | 5 | 1109 |
Gene Symbol: Abcg3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037205.1 | NP_001032282.1 | 5 | 487 | ||
GenBank mRNA | BC100073.1 | - | 5 | 487 | 7372627 |
Gene Symbol: LOC103690063 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769965.1 | XP_008768187.1 | 5 | 1059 |

More Information
Other Species Targeted

Gene Ontology Categories:
