siRNA Details
Gene Symbol: Abcg3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004076.2 | NP_001004076.2 | 9 | 1090 | ||
XM_006250626.2 | XP_006250688.1 | 9 | 1139 | |||
XM_008770002.1 | XP_008768224.1 | 9 | 3480 | |||
XM_008770003.1 | XP_008768225.1 | 10 | 3360 | |||
GenBank mRNA | AY688113.1 | - | 8 | 1017 | ||
BC098896.1 | - | 9 | 1090 | 7100556 |
Gene Symbol: Abcg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128311.1 | NP_001121783.1 | 8 | 1017 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
