siRNA Details
Gene Symbol: Lrrc36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004088.3 | NP_001004088.3 | 8 | 903 | ||
XM_006255503.1 | XP_006255565.1 | 8 | 891 | |||
XM_006255504.1 | XP_006255566.1 | 8 | 891 | |||
XM_006255505.1 | XP_006255567.1 | 8 | 891 | |||
XM_006255508.2 | XP_006255570.1 | 6 | 1056 | |||
XM_006255509.2 | XP_006255571.1 | 8 | 832 | |||
GenBank mRNA | AY248698.1 | - | 7 | 874 |

More Information
Other Species Targeted

