siRNA Details
Gene Symbol: Lrrc36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004088.3 | NP_001004088.3 | 12 | 1848 | ||
XM_006255503.1 | XP_006255565.1 | 12 | 1830 | |||
XM_006255504.1 | XP_006255566.1 | 12 | 1830 | |||
XM_006255505.1 | XP_006255567.1 | 12 | 1695 | |||
XM_006255507.1 | XP_006255569.1 | 11 | 1383 | |||
XM_006255508.2 | XP_006255570.1 | 10 | 1995 | |||
XM_006255509.2 | XP_006255571.1 | 12 | 1771 | |||
GenBank mRNA | AY248698.1 | - | 11 | 1806 |

More Information
Other Species Targeted

