siRNA Details
Gene Symbol: ARHGAP11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286479.1 | NP_001273408.1 | 12 | 4271 | ||
NM_001286480.1 | NP_001273409.1 | 13 | 4393 | |||
NM_014783.4 | NP_055598.1 | 12 | 5012 | |||
GenBank mRNA | D87717.2 | - | 12 | 5012 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
