siRNA Details
Gene Symbol: NPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005251479.4 | XP_005251536.1 | 1 | 3539 | ||
XM_011517890.2 | XP_011516192.1 | 1 | 3539 | |||
XM_011517891.2 | XP_011516193.1 | 1 | 3539 | |||
XM_011517892.2 | XP_011516194.1 | 1 | 3539 | |||
XM_011517893.2 | XP_011516195.1 | 1 | 3539 | |||
XM_017014745.1 | XP_016870234.1 | 1 | 3539 | |||
XM_017014746.1 | XP_016870235.1 | 1 | 3539 | |||
XM_017014747.1 | XP_016870236.1 | 1 | 3539 | |||
XM_017014748.1 | XP_016870237.1 | 1 | 3539 | |||
XM_017014749.1 | XP_016870238.1 | 1 | 3539 | |||
XM_017014750.1 | XP_016870239.1 | 1 | 3539 |
Gene Symbol: RGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | D87447.1 | - | 9 | 4871 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
