siRNA Details
Gene Symbol: B130024G19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK045070.1 | - | 8 | 921 | ||
BC070425.1 | - | 8 | 1152 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_001004183.1 | - | 921 | Chr. - - [-] on Build |
