siRNA Details
Gene Symbol: BC048562 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004192.1 | NP_001004192.1 | 2, 3 | 223 | ||
GenBank mRNA | AK029636.1 | - | 2, 3 | 224 | ||
BC048562.1 | - | 2, 3 | 223 | 6703719 |
