siRNA Details
Gene Symbol: Nqo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004214.1 | NP_001004214.1 | 7 | 828 | ||
XM_006253867.2 | XP_006253929.1 | 7 | 1079 | |||
XM_006253868.2 | XP_006253930.1 | 6 | 837 | |||
GenBank mRNA | BC079157.1 | - | 7 | 828 | 7128990 |
