siRNA Details
Gene Symbol: Mkrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004233.1 | NP_001004233.1 | 5 | 869 | ||
XM_006236301.2 | XP_006236363.1 | 5 | 1069 | |||
XM_006236302.2 | XP_006236364.1 | 4 | 941 | |||
XM_006236303.2 | XP_006236365.1 | 5 | 1025 | |||
XM_006236304.2 | XP_006236366.1 | 5 | 885 | |||
GenBank mRNA | BC079407.1 | - | 5 | 869 | 7112867 | |
FQ212797.1 | - | 3 | 674 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_018810, AK215701, AF192785, AB026986, AK005137, and others.
Gene Ontology Categories:
