siRNA Details
Gene Symbol: Tmem30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004248.1 | NP_001004248.1 | 5 | 996 | ||
XM_006243435.2 | XP_006243497.1 | 6 | 1106 | |||
GenBank mRNA | BC079203.1 | - | 5 | 996 | 7131766 | |
FQ209432.1 | - | 2 | 186 |
