siRNA Details
Gene Symbol: Uqcrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004250.2 | NP_001004250.1 | 13 | 1555 | ||
GenBank mRNA | BC078923.1 | - | 13 | 1557 | 7106265 | |
FQ210217.1 | - | 6 | 674 | |||
FQ213124.1 | - | 13 | 1566 | |||
FQ215592.1 | - | 9 | 1039 | |||
FQ216746.1 | - | 13 | 1559 | |||
FQ224157.1 | - | 13 | 1563 | |||
FQ229052.1 | - | 13 | 1582 | |||
FQ231552.1 | - | 13 | 1579 |
