siRNA Details
Gene Symbol: Syap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004253.1 | NP_001004253.1 | 3 | 539 | ||
XM_006256889.2 | XP_006256951.1 | 3 | 552 | |||
GenBank mRNA | BC079116.1 | - | 3 | 539 | 7126243 | |
FQ219851.1 | - | 2 | 117 |
