siRNA Details
Gene Symbol: Sirt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004256.1 | NP_001004256.1 | 8 | 1042 | ||
XM_006253801.1 | XP_006253863.1 | 9 | 1226 | |||
XM_006253802.1 | XP_006253864.1 | 9 | 1104 | |||
XM_006253803.2 | XP_006253865.1 | 9 | 1098 | |||
XM_006253804.2 | XP_006253866.1 | 9 | 1049 | |||
GenBank mRNA | BC078958.1 | - | 8 | 1042 | 7110841 |
